Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- acdh-10
- Entrez Gene ID
-
24104866
- Gene Name
- Probable medium-chain specific acyl-CoA dehydrogenase 10, mitochondrial
- Gene Aliases
-
CELE_T08G2.3
- Chromosome Location
-
Chr.X: 17213144 - 17214903 on Build WBcel235
- Gene Symbol
- acdh-7
- Entrez Gene ID
-
181758
- Gene Name
- Acyl CoA DeHydrogenase
- Gene Aliases
-
CELE_T25G12.5
- Chromosome Location
-
Chr.X: 17213144 - 17217105 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078388.5 |
NP_510789.1 |
6 - 7 |
2364 |
81 |
View Full Product Details