Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptr-5
Entrez Gene ID
181764
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_C53C11.3
Chromosome Location
Chr.X: 17318584 - 17326625 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078409.8 NP_510810.3 11 - 12 1425 59

View Full Product Details