Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sup-10
Entrez Gene ID
181784
Gene Name
Putative potassium channel regulatory protein sup-10
Gene Aliases
CELE_R09G11.1
Chromosome Location
Chr.X: 17528344 - 17531985 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078434.7 NP_510835.1 4 - 5 635 60
GenBank mRNA U43891.1 - 4 - 5 585 60

View Full Product Details