Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-9
Entrez Gene ID
181786
Gene Name
Galectin
Gene Aliases
CELE_C16H3.2
Chromosome Location
Chr.X: 17600293 - 17601503 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078443.8 NP_510844.1 3 - 4 456 98
GenBank mRNA AB038500.1 - 3 - 4 333 98

View Full Product Details