Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cgt-2
Entrez Gene ID
181790
Gene Name
Ceramide glucosyltransferase 2
Gene Aliases
CELE_F20B4.6
Chromosome Location
Chr.X: 17695232 - 17701744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078456.5 NP_510857.2 3 - 4 580 119
GenBank mRNA AF364401.1 - 3 - 4 584 119

View Full Product Details