Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hrpf-1
Entrez Gene ID
172395
Gene Name
protein-coding
Gene Aliases
CELE_W02D3.11
Chromosome Location
Chr.I: 6753219 - 6756354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372997.1 NP_001359979.1 2 - 3 214 111
NM_170888.6 NP_740877.1 2 - 3 220 111

View Full Product Details