Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aex-1
Entrez Gene ID
172519
Gene Name
protein-coding
Gene Aliases
CELE_D2030.10
Chromosome Location
Chr.I: 7602307 - 7608245 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170901.4 NP_740891.1 17 - 18 2765 68
GenBank mRNA AF324832.1 - 17 - 18 2761 68

View Full Product Details