Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aex-1
Entrez Gene ID
172519
Gene Name
protein-coding
Gene Aliases
CELE_D2030.10
Chromosome Location
Chr.I: 7602307 - 7608245 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025903.4 NP_001021074.1 3 - 4 227 147
NM_170901.4 NP_740891.1 3 - 4 244 147
GenBank mRNA AF324832.1 - 3 - 4 240 147

View Full Product Details