Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps-20
Entrez Gene ID
173309
Gene Name
Ribosomal_S10 domain-containing protein
Gene Aliases
CELE_Y105E8A.16
Chromosome Location
Chr.I: 14468218 - 14469103 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170948.7 NP_740944.1 2 - 3 190 57

View Full Product Details