Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-27
Entrez Gene ID
174121
Gene Name
protein-coding
Gene Aliases
CELE_C04A2.3
Chromosome Location
Chr.II: 6835008 - 6847908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276731.3 NP_001263660.1 6 - 7 2444 82
NM_001313565.3 NP_001300494.1 12 - 12 3161 82
NM_001313566.3 NP_001300495.1 12 - 12 3158 82
NM_001313568.3 NP_001300497.1 11 - 12 3065 82
NM_001313569.3 NP_001300498.1 11 - 12 3059 82
NM_001313570.3 NP_001300499.1 11 - 12 3056 82
NM_001313571.3 NP_001300500.1 11 - 12 3050 82
NM_001313572.3 NP_001300501.1 2 - 3 1622 82
NM_001313575.3 NP_001300504.1 9 - 10 2729 82
NM_001381461.1 NP_001368508.1 12 - 13 3152 82
NM_001381462.1 NP_001368509.1 9 - 10 2738 82
NM_001381463.1 NP_001367420.1 12 - 13 3231 82
NM_001381464.1 NP_001368510.1 6 - 7 2447 82
NM_001381465.1 NP_001368511.1 6 - 7 2438 82
NM_001383871.1 12 - 13 3211 82
NM_001383872.1 11 - 12 3188 82
NM_001383873.1 12 - 13 3227 82
NM_171011.4 NP_741012.1 12 - 13 3227 82

View Full Product Details