Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-38
Entrez Gene ID
174136
Gene Name
protein-coding
Gene Aliases
CELE_F22D3.1
Chromosome Location
Chr.II: 6951526 - 6955409 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351905.4 NP_001338814.1 5 - 5 1019 89
NM_001351913.2 NP_001338820.1 5 - 6 901 89
NM_001351914.2 NP_001338821.1 5 - 6 1010 89
NM_001381471.2 NP_001368648.1 5 - 6 884 89
NM_001381472.2 NP_001368657.1 5 - 6 918 89
NM_001381477.2 NP_001367332.1 5 - 6 870 89
NM_001381478.2 NP_001368653.1 4 - 5 837 89
NM_001383875.1 5 - 6 1019 89

View Full Product Details