Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-38
Entrez Gene ID
174136
Gene Name
protein-coding
Gene Aliases
CELE_F22D3.1
Chromosome Location
Chr.II: 6951526 - 6955409 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351905.4 NP_001338814.1 8 - 8 1836 74
NM_001351913.2 NP_001338820.1 8 - 9 1718 74
NM_001381471.2 NP_001368648.1 8 - 9 1695 74
NM_001381473.2 NP_001368649.1 7 - 8 1630 74
NM_001381475.2 NP_001368651.1 8 - 9 1710 74
NM_001381477.2 NP_001367332.1 8 - 9 1681 74
NM_001381479.1 NP_001368654.1 7 - 8 1622 74
NM_171852.2 NP_741018.1 8 - 9 1680 74

View Full Product Details