Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-21
Entrez Gene ID
174232
Gene Name
protein-coding
Gene Aliases
CELE_R07G3.3
Chromosome Location
Chr.II: 7599657 - 7608016 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373741.2 NP_001360471.1 13 - 14 4256 127
NM_001373742.2 NP_001360472.1 13 - 14 4256 127
NM_001373743.2 NP_001360024.1 13 - 14 4256 127
NM_001373744.2 NP_001360023.1 13 - 14 4258 127

View Full Product Details