Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptb-1
Entrez Gene ID
174647
Gene Name
protein-coding
Gene Aliases
CELE_D2089.4
Chromosome Location
Chr.II: 10675602 - 10690408 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171035.6 NP_741041.1 8 - 9 1077 92
NM_171036.6 NP_741042.1 3 - 4 650 92
GenBank mRNA LC026472.1 - 7 - 8 1021 92
LC026473.1 - 3 - 4 535 92

View Full Product Details