Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- gsto-3
- Entrez Gene ID
-
175196
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_K10F12.4
- Chromosome Location
-
Chr.III: 378004 - 381298 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171061.1 |
NP_741069.1 |
3 - 4 |
587 |
112 |
- Gene Symbol
- gei-4
- Entrez Gene ID
-
175195
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_W07B3.2
- Chromosome Location
-
Chr.III: 344306 - 418744 on Build WBcel235
View Full Product Details