Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab-18
Entrez Gene ID
175272
Gene Name
protein-coding
Gene Aliases
CELE_Y92C3B.3
Chromosome Location
Chr.III: 1078138 - 1080775 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171081.6 NP_741092.1 4 - 5 527 129

View Full Product Details