Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ptp-1
Entrez Gene ID
175463
Gene Name
protein-coding
Gene Aliases
CELE_C48D5.2
Chromosome Location
Chr.III: 3590722 - 3611971 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171099.5 NP_741112.1 1 - 2 28 62

View Full Product Details