Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
skpt-1
Entrez Gene ID
175749
Gene Name
protein-coding
Gene Aliases
CELE_F48E8.7
Chromosome Location
Chr.III: 5446511 - 5448419 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171121.5 NP_741136.1 1 - 2 228 81

View Full Product Details