Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
efk-1
Entrez Gene ID
175871
Gene Name
protein-coding
Gene Aliases
CELE_F42A10.4
Chromosome Location
Chr.III: 6164878 - 6168547 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027401.3 NP_001022572.1 4 - 5 1909 73
GenBank mRNA U93846.1 - 4 - 5 1892 73

View Full Product Details