Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gar-2
Entrez Gene ID
175893
Gene Name
protein-coding
Gene Aliases
CELE_F47D12.1
Chromosome Location
Chr.III: 6304490 - 6320872 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027422.4 NP_001022593.1 8 - 9 1405 141
NM_001027423.6 NP_001022594.1 8 - 9 1417 141
NM_001027426.4 NP_001022597.1 8 - 9 1411 141
NM_001379757.2 NP_001368076.1 10 - 11 1541 141
GenBank mRNA AF272738.1 - 8 - 9 1362 141
AY053364.1 - 8 - 9 1362 141
AY053365.1 - 8 - 9 1362 141

View Full Product Details