Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mel-32
Entrez Gene ID
175915
Gene Name
protein-coding
Gene Aliases
CELE_C05D11.11
Chromosome Location
Chr.III: 6440145 - 6443980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379763.1 NP_001367440.1 4 - 5 1650 92
NM_171169.6 NP_741197.1 3 - 4 529 92

View Full Product Details