Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nuo-4
Entrez Gene ID
176001
Gene Name
protein-coding
Gene Aliases
CELE_K04G7.4
Chromosome Location
Chr.III: 7156026 - 7158809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379784.1 NP_001367335.1 2 - 3 601 124
NM_171184.5 NP_741215.1 2 - 3 608 124

View Full Product Details