Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gei-8
Entrez Gene ID
176145
Gene Name
protein-coding
Gene Aliases
CELE_C14B9.6
Chromosome Location
Chr.III: 8111168 - 8126222 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066372.7 NP_498773.3 16 - 17 5006 63
NM_171207.8 NP_741245.3 15 - 16 4757 63

View Full Product Details