The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001028392.7 | NP_001023563.1 | 4 - 5 | 403 | 92 |
NM_001307684.3 | NP_001294613.1 | 2 - 3 | 145 | 92 | |
GenBank mRNA | AF239999.2 | - | 3 - 4 | 353 | 92 |