Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ser-6
Entrez Gene ID
259560
Gene Name
protein-coding
Gene Aliases
CELE_Y54G2A.35
Chromosome Location
Chr.IV: 2960098 - 2964401 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171297.4 NP_741350.1 4 - 5 874 97

View Full Product Details