Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-33
Entrez Gene ID
177112
Gene Name
protein-coding
Gene Aliases
CELE_Y37E11C.1
Chromosome Location
Chr.IV: 3516886 - 3526602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038349.4 NP_001033438.1 5 - 6 540 68
GenBank mRNA Z14146.1 - - 605 68

View Full Product Details