Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
skn-1
Entrez Gene ID
177343
Gene Name
protein-coding
Gene Aliases
CELE_T19E7.2
Chromosome Location
Chr.IV: 5651039 - 5660384 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380237.1 NP_001367379.1 3 - 4 897 74
NM_171345.6 NP_741404.1 5 - 6 1167 74

View Full Product Details