Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mut-2
Entrez Gene ID
187005
Gene Name
protein-coding
Gene Aliases
CELE_K04F10.6
Chromosome Location
Chr.I: 6356614 - 6358411 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059433.8 NP_491834.1 6 - 7 1025 105

View Full Product Details