Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-11
Entrez Gene ID
191599
Gene Name
AcetylCholine Receptor
Gene Aliases
CELE_D2092.3
Chromosome Location
Chr.I: 6621624 - 6624918 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059505.2 NP_491906.1 8 - 9 1176 88

View Full Product Details