Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nekl-2
Entrez Gene ID
191199
Gene Name
Serine/threonine-protein kinase nekl-2
Gene Aliases
CELE_ZC581.1
Chromosome Location
Chr.I: 6663050 - 6665160 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059513.5 NP_491914.1 7 - 8 1021 113

View Full Product Details