Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bbs-9
Entrez Gene ID
183572
Gene Name
Protein pthb1 homolog
Gene Aliases
CELE_C48B6.8
Chromosome Location
Chr.I: 6917700 - 6922973 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059572.6 NP_491973.4 9 - 10 1329 132

View Full Product Details