Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nduf-6
- Entrez Gene ID
-
184827
- Gene Name
- Probable NADH dehydrogenase [ubiquinone] iron-sulfur protein 6, mitochondrial
- Gene Aliases
-
CELE_F22D6.4
- Chromosome Location
-
Chr.I: 7087458 - 7091365 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059606.9 |
NP_492007.1 |
3 - 4 |
358 |
69 |
View Full Product Details