Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ekl-1
Entrez Gene ID
172444
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F22D6.6
Chromosome Location
Chr.I: 7095486 - 7098406 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059608.5 NP_492009.1 1 - 2 138 83

View Full Product Details