Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-14
Entrez Gene ID
172450
Gene Name
RUN domain-containing protein
Gene Aliases
CELE_K10D3.2
Chromosome Location
Chr.I: 7122614 - 7128427 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059617.6 NP_492018.1 6 - 7 1544 101
GenBank mRNA AB000913.1 - 6 - 7 1562 101

View Full Product Details