Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-15
Entrez Gene ID
172470
Gene Name
protein-coding
Gene Aliases
CELE_R06C7.9
Chromosome Location
Chr.I: 7255380 - 7258702 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129030.3 NP_001122502.1 6 - 7 1155 112
NM_059651.6 NP_492052.2 6 - 7 1024 112

View Full Product Details