Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-53
Entrez Gene ID
184430
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_F13G3.2
Chromosome Location
Chr.I: 7294038 - 7295794 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059660.2 NP_492061.1 5 - 6 827 69

View Full Product Details