Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mif-3
- Entrez Gene ID
-
172481
- Gene Name
- MIF-like protein mif-3
- Gene Aliases
-
CELE_F13G3.9
- Chromosome Location
-
Chr.I: 7312797 - 7313525 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_059668.4 |
NP_492069.1 |
1 - 2 |
56 |
82 |
- Gene Symbol
- F13G3.10
- Entrez Gene ID
-
3565846
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F13G3.10
- Chromosome Location
-
Chr.I: 7312793 - 7314279 on Build WBcel235
View Full Product Details