Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ego-1
Entrez Gene ID
172524
Gene Name
Enhancer of Glp-One (glp-1)
Gene Aliases
CELE_F26A3.3
Chromosome Location
Chr.I: 7650341 - 7656338 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059731.6 NP_492132.1 8 - 9 2665 103
GenBank mRNA AF159143.1 - 8 - 9 2663 103

View Full Product Details