Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-8
Entrez Gene ID
191617
Gene Name
protein-coding
Gene Aliases
CELE_ZK265.4
Chromosome Location
Chr.I: 8245505 - 8250948 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318276.4 NP_001305205.1 5 - 5 571 121
NM_001318277.3 NP_001305206.1 2 - 2 141 121
NM_059845.4 NP_492246.2 4 - 5 595 121

View Full Product Details