Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsd-6
Entrez Gene ID
172621
Gene Name
Tudor domain-containing protein
Gene Aliases
CELE_F16D3.2
Chromosome Location
Chr.I: 8358248 - 8363858 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059868.9 NP_492269.2 2 - 3 410 87

View Full Product Details