Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rskn-1
Entrez Gene ID
172647
Gene Name
protein-coding
Gene Aliases
CELE_T01H8.1
Chromosome Location
Chr.I: 8573852 - 8580205 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026431.3 NP_001021602.1 9 - 10 1824 89
NM_001026432.4 NP_001021603.1 10 - 11 1837 89
NM_001129032.4 NP_001122504.1 10 - 11 1843 89
NM_001373180.3 NP_001359590.1 11 - 12 2059 89
NM_059918.4 NP_492319.2 10 - 11 1887 89

View Full Product Details