Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snf-2
Entrez Gene ID
172701
Gene Name
Transporter
Gene Aliases
CELE_F55H12.1
Chromosome Location
Chr.I: 8871791 - 8875257 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059995.5 NP_492396.2 5 - 6 1698 90

View Full Product Details