Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smn-1
Entrez Gene ID
172783
Gene Name
protein-coding
Gene Aliases
CELE_C41G7.1
Chromosome Location
Chr.I: 9511174 - 9512149 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381073.1 NP_001366985.1 4 - 5 409 103
NM_001383633.1 4 - 5 458 103
GenBank mRNA AF156887.1 - 4 - 5 434 103
Y17255.1 - - 385 103

View Full Product Details