Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lem-3
Entrez Gene ID
172792
Gene Name
Ankyrin repeat and LEM domain-containing protein 1 homolog
Gene Aliases
CELE_F42H11.2
Chromosome Location
Chr.I: 9578526 - 9581267 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060138.4 NP_492539.2 2 - 3 334 71

View Full Product Details