Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ile-1
Entrez Gene ID
172799
Gene Name
L-type lectin-like domain-containing protein
Gene Aliases
CELE_K07A1.8
Chromosome Location
Chr.I: 9609497 - 9611534 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060147.6 NP_492548.1 1 - 2 80 88
GenBank mRNA DQ384615.1 - 1 - 2 78 88

View Full Product Details