Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gfi-2
Entrez Gene ID
172825
Gene Name
protein-coding
Gene Aliases
CELE_K02A11.1
Chromosome Location
Chr.I: 9737816 - 9744146 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381136.2 NP_001366793.1 5 - 6 1383 66

View Full Product Details