Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-4
Entrez Gene ID
172834
Gene Name
protein-coding
Gene Aliases
CELE_F35C12.2
Chromosome Location
Chr.I: 9808904 - 9813075 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047198.5 NP_001040663.1 1 - 2 211 138

View Full Product Details