Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-62
Entrez Gene ID
172836
Gene Name
protein-coding
Gene Aliases
CELE_Y67A6A.2
Chromosome Location
Chr.I: 9839598 - 9844809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373584.2 NP_001360798.1 1 - 2 260 77
NM_060206.7 NP_492607.1 1 - 2 262 77

View Full Product Details