Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zig-1
Entrez Gene ID
192086
Gene Name
Zwei Ig domain protein zig-1
Gene Aliases
CELE_K10C3.3
Chromosome Location
Chr.I: 9847406 - 9850082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060207.6 NP_492608.2 6 - 7 693 145
GenBank mRNA AF456248.1 - 6 - 7 692 145

View Full Product Details