Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mom-4
Entrez Gene ID
172842
Gene Name
Mitogen-activated protein kinase kinase kinase mom-4
Gene Aliases
CELE_F52F12.3
Chromosome Location
Chr.I: 9895678 - 9900817 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060219.9 NP_492620.1 8 - 9 1463 106
GenBank mRNA AF143242.1 - 8 - 9 1454 106
AF145377.1 - 8 - 9 1245 106

View Full Product Details